Assay Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181058.1 | NP_851401.1 | 4 - 5 | 324 | 78 | |
| GenBank mRNA | AF194030.1 | - | 2 - 3 | 161 | 78 | |
| AK005028.2 | - | 4 - 5 | 325 | 78 | ||
| AK078664.1 | - | 308 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF194030.1, AK005028.2, NM_181058.1 transcripts.
Set Membership: |
|

Zbtb20
Genomic Map