Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290644.1 | NP_001277573.1 | 1 - 2 | 343 | 84 | |
| GenBank mRNA | AK033954.1 | - | 1 - 2 | 266 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033954.1, NM_001290644.1 transcripts.
Set Membership: |
|

Rp2h
Genomic Map