Assay Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195284.1 | NP_001182213.1 | 9 - 10 | 1052 | 89 | |
| GenBank mRNA | BB626514.1 | - | 2 - 3 | 179 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fsd1l
Genomic Map