Assay Details
| Gene Symbol: Hhatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029095.2 | NP_083371.2 | 3 - 4 | 432 | 61 | |
| NR_027967.1 | 3 - 4 | 312 | 61 | |||
| GenBank mRNA | AK003605.1 | - | 3 - 4 | 309 | 61 | |
| AK157924.1 | - | 3 - 4 | 230 | 61 | ||
| BC024464.1 | - | 3 - 4 | 288 | 4975643 | 61 | |
| BY703867.1 | - | 3 - 4 | 309 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hhatl
Genomic Map