Assay Details
| Gene Symbol: Hhatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029095.2 | NP_083371.2 | 5 - 6 | 744 | 86 | |
| NR_027967.1 | 5 - 6 | 624 | 86 | |||
| GenBank mRNA | AK157924.1 | - | 5 - 6 | 542 | 86 | |
| BC024464.1 | - | 5 - 6 | 600 | 4975643 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hhatl
Genomic Map