Assay Details
| Gene Symbol: Nipsnap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025623.2 | NP_079899.1 | 4 - 5 | 640 | 89 | |
| GenBank mRNA | AF353623.1 | - | 4 - 5 | 629 | 89 | |
| AK004458.1 | - | 4 - 5 | 639 | 89 | ||
| AK008786.1 | - | 4 - 5 | 636 | 89 | ||
| AK012480.1 | - | 4 - 5 | 640 | 89 | ||
| BC019505.1 | - | 4 - 5 | 625 | 4164310 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipsnap3b
Genomic Map