Assay Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183029.2 | NP_898850.2 | 5 - 6 | 735 | 83 | |
| GenBank mRNA | AK044984.1 | - | 4 - 5 | 251 | 83 | |
| AK049196.1 | - | 5 - 6 | 432 | 83 | ||
| AK077404.1 | - | 5 - 6 | 415 | 83 | ||
| AK170531.1 | - | 4 - 5 | 239 | 83 | ||
| AY531659.1 | - | 5 - 6 | 399 | 83 | ||
| BC023758.1 | - | 5 - 6 | 433 | 83 | ||
| BC054552.1 | - | 5 - 6 | 455 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp2
Genomic Map