Assay Details
| Gene Symbol: Gm19619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040428.1 | 2 - 3 | 276 | 113 | ||
| GenBank mRNA | AK050091.1 | - | 2 - 3 | 277 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | amphiregulin | Areg |
AR, Mcub, Sdgf |
-131.3kb | Anti-sense | Chr. 5 - 91139615 - 91148432 on NCBI Build GRCm38 |
| Downstream |
betacellulin, epidermal growth factor family member |
Btc | Bcn | 75.6kb | Sense | Chr. 5 - 91357261 - 91402994 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1400000269.rn
Set Membership: |
|

Gm19619
Genomic Map