Assay Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002926.3 | 1 - 2 | 45 | 83 | ||
| GenBank mRNA | AK131857.1 | - | 1 - 2 | 46 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2a
Genomic Map