Assay Details
| Gene Symbol: Gm39182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038453.1 | - | 105 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence AW046200 | AW046200 | -36.3kb | Sense | Chr. 8 - 57651753 - 57663430 on NCBI Build GRCm38 | |
| Downstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 6 | Galntl6 |
1700021K10Rik, 4930431L04Rik, A830023L05 |
72.4kb | Anti-sense | Chr. 8 - 57772489 - 58911627 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm39182
Genomic Map