Assay Details
Gene Symbol: 9530051G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040272.1 | 1 - 2 | 63 | 78 | ||
GenBank mRNA | AK035460.1 | - | 1 - 2 | 64 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | forkhead box R2 | Foxr2 |
EG436240, Foxn6 |
Sense | Chr. X - 153016795 - 153132861 on NCBI Build GRCm38 | |
Downstream | Ras-related GTP binding B | Rragb | 101.0kb | Sense | Chr. X - 153139796 - 153190192 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
