Assay Details
| Gene Symbol: C730014E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050351.1 | - | 1 - 2 | 117 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
adiponectin, C1Q and collagen domain containing |
Adipoq |
30kDa, APN, Acdc, Acrp30, Ad, GBP28, adipo, apM1 |
-34.2kb | Sense | Chr. 16 - 23146536 - 23157968 on NCBI Build GRCm38 |
| Downstream | cDNA sequence BC106179 | BC106179 | 26.0kb | Anti-sense | Chr. 16 - 23219010 - 23225884 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

C730014E05Rik
Genomic Map