Assay Details
| Gene Symbol: Gm12409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046068.1 | 1 - 2 | 80 | 61 | ||
| GenBank mRNA | AK041524.1 | - | 1 - 2 | 81 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 829 | Gm829 | -39.3kb | Anti-sense | Chr. 4 - 45683589 - 45723706 on NCBI Build GRCm38 | |
| Downstream |
aldehyde dehydrogenase 1 family, member B1 |
Aldh1b1 | 2700007F14Rik | 34.8kb | Anti-sense | Chr. 4 - 45799022 - 45804608 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12409
Genomic Map