Assay Details
| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079255.1 | - | 314 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A730085K08 gene | A730085K08Rik | -178.1kb | Anti-sense | Chr. 9 - 122382022 - 122398801 on NCBI Build GRCm38 | |
| Downstream | microRNA 138-1 | Mir138-1 |
Mirn138, Mirn138-1, mir-138-1 |
30.7kb | Sense | Chr. 9 - 122682876 - 122682974 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

9530059O14Rik
Genomic Map