Assay Details
Gene Symbol: Fhad1os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040672.1 | 1 - 2 | 47 | 65 | ||
GenBank mRNA | AK079199.1 | - | 1 - 2 | 48 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 42338 |
Gm42338 | -67.1kb | Sense | Chr. 4 - 141909876 - 141915922 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930455G09 gene | 4930455G09Rik | 34.3kb | Sense | Chr. 4 - 142017669 - 142029000 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
