Assay Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163006.2 | NP_001156478.1 | 3 - 4 | 344 | 71 | |
| NR_033745.1 | 3 - 4 | 402 | 71 | |||
| GenBank mRNA | AA023452.1 | - | 3 - 4 | 359 | 71 | |
| AK004466.1 | - | 3 - 4 | 236 | 71 | ||
| AK008362.1 | - | 3 - 4 | 228 | 71 | ||
| AK008449.1 | - | 3 - 4 | 228 | 71 | ||
| AK009189.1 | - | 3 - 4 | 227 | 71 | ||
| AK009528.1 | - | 3 - 4 | 270 | 71 | ||
| AK011553.1 | - | 3 - 4 | 227 | 71 | ||
| AK030343.1 | - | 3 - 4 | 297 | 71 | ||
| AK152509.1 | - | 3 - 4 | 270 | 71 | ||
| AK153063.1 | - | 3 - 4 | 269 | 71 | ||
| AK159039.1 | - | 3 - 4 | 223 | 71 | ||
| AK159100.1 | - | 3 - 4 | 270 | 71 | ||
| AK167818.1 | - | 3 - 4 | 269 | 71 | ||
| BC055791.1 | - | 3 - 4 | 261 | 6410794 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349883 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123962 | AK159039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123962 | AK159100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123962 | AK153063.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123962 | AK167818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123962 | BC055791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK008170.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK008207.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK008243.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK079604.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK148438.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | AK165381.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | BC048371.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | NM_001081495.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916283 | NM_025988.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30088 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55555 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136510 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158457 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138142 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123962 | AK159039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123962 | AK159100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123962 | AK153063.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123962 | AK167818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123962 | BC055791.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136510 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138142 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05424180 | - | - |
| Whole Mouse Genome G4122A | A_52_P571403 | AK159039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P571403 | AK159100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P571403 | AK153063.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P571403 | AK167818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P571403 | BC055791.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | AK030343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | NM_001163006.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486639 | NR_033745.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138142 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136510 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03547436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04004940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04024554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03722491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03547434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03716638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03713003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03659560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03722495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04005890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03306565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03337627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03641937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03872651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03785789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03872652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03370082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03713001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03360563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03782064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03712999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03577398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03659559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03641942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03634463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03370075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03722497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03872656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03850688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04033194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03275705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03426478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03370085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03275715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03755135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03547438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03540216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03872650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03950101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03716642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04004942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03926669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03417502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03275711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23349913 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517686 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517684 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455126_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436001_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433616_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517682 | - | - |
| Mouse Genome 430 2.0 Array | 1455126_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436001_at | - | - |
| Mouse Genome 430 2.0 Array | 1433616_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1436001_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433616_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455126_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433771_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433771_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400031674.mm.1 | TC0400003842.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031674.mm.1 | NR_033745.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031674.mm.1 | NR_033745.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031668.mm.1 | TC0400003842.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031668.mm.1 | NM_001163006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031668.mm.1 | NM_001163006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031668.mm.1 | NR_033745.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031668.mm.1 | NR_033745.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031669.mm.1 | TC0400003842.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031669.mm.1 | NM_001163006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031669.mm.1 | NM_001163006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031669.mm.1 | NR_033745.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031669.mm.1 | NR_033745.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031665.mm.1 | TC0400003842.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031665.mm.1 | NM_001163006.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031665.mm.1 | NM_001163006.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031665.mm.1 | NR_033745.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031665.mm.1 | NR_033745.1 | Refseq |
| Mouse Expression Set 430 | 1433616_a_at | - | - |
| Mouse Expression Set 430 | 1433616_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455126_PM_x_at | - | - |
| Mouse Expression Set 430 | 1455126_x_at | - | - |
| Mouse Expression Set 430 | 1436001_at | - | - |
| Mouse Expression Set 430 | 1436001_PM_at | - | - |
| Murine Genome U74v2 Set | 96261_PM_at | - | - |
| Murine Genome U74v2 Set | 96261_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433616_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433616_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455126_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455126_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455126_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433616_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455126_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433616_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436001_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436001_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 718044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103710014 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2440183 | - | - |

Minos1
Genomic Map