Assay Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025290.3 | NP_079566.1 | 1 - 2 | 152 | 69 | |
| GenBank mRNA | AB006535.1 | - | 1 - 2 | 140 | 69 | |
| AK005739.1 | - | 1 - 2 | 104 | 69 | ||
| BC049584.1 | - | 1 - 2 | 152 | 6744132 | 69 | |
| DQ141549.1 | - | 1 - 2 | 51 | 69 | ||
| DQ141550.1 | - | 1 - 2 | 51 | 69 | ||
| DQ141551.1 | - | 1 - 2 | 51 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsph1
Genomic Map