Assay Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025290.3 | NP_079566.1 | 2 - 3 | 268 | 108 | |
| GenBank mRNA | AK005739.1 | - | 2 - 3 | 220 | 108 | |
| BC049584.1 | - | 2 - 3 | 268 | 6744132 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rsph1
Genomic Map