Assay Details
| Gene Symbol: Slc37a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242427.1 | NP_001229356.1 | 13 - 14 | 1980 | 65 | |
| NM_153062.2 | NP_694702.1 | 13 - 14 | 1560 | 65 | ||
| GenBank mRNA | AK033656.1 | - | 13 - 14 | 1449 | 65 | |
| AK143636.1 | - | 13 - 14 | 1986 | 65 | ||
| AK153706.1 | - | 13 - 14 | 1983 | 65 | ||
| AK163990.1 | - | 12 - 13 | 1162 | 65 | ||
| BC027294.1 | - | 13 - 14 | 1560 | 3985469 | 65 | |
| BC029208.1 | - | 5 - 6 | 380 | 65 | ||
More Information
Set Membership: |
|

Slc37a1
Genomic Map