Assay Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046122.1 | - | 8 - 9 | 1768 | 121 | |
| AK193923.1 | - | 3 - 4 | 338 | 121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grik1| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046122.1 | - | 8 - 9 | 1768 | 121 | |
| AK193923.1 | - | 3 - 4 | 338 | 121 | ||
Set Membership: |
|