| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_001164612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_001164612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_001164613.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_001164613.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_172613.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
NM_172613.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011920.mm.1 |
TC1600001443.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_001164612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_001164612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_001164613.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_001164613.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_172613.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
NM_172613.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011921.mm.1 |
TC1600001443.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_001164612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_001164612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_001164613.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_001164613.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_172613.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
NM_172613.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600011919.mm.1 |
TC1600001443.mm.1 |
MTA TC |