Assay Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028684.1 | - | 1 - 2 | 143 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028684.1 transcripts.
Set Membership: |
|

Psd3
Genomic Map