Assay Details
| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177505.1 | NP_001170976.1 | 1 - 2 | 483 | 110 | |
| GenBank mRNA | BC042569.1 | - | 2 - 3 | 764 | 110 | |
| BC119591.1 | - | 1 - 2 | 483 | 40058904 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan7
Genomic Map