Assay Details
| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177505.1 | NP_001170976.1 | 3 - 4 | 701 | 88 | |
| GenBank mRNA | BC119591.1 | - | 3 - 4 | 701 | 40058904 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan7
Genomic Map