Assay Details
| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177505.1 | NP_001170976.1 | 4 - 5 | 796 | 76 | |
| GenBank mRNA | BC119591.1 | - | 4 - 5 | 796 | 40058904 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891988 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891893 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142326 | BC042569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142326 | AK041109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034270 | BC119591.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034270 | NM_001177505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034270 | AK163886.1 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891988 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26891990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07696409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07778537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07459035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08014982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07778539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07867653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07781459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07696410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07700245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26891913 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10590480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590484 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10590479 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900013835.mm.1 | NM_001177505.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013835.mm.1 | NM_001177505.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013835.mm.1 | TC0900001647.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1442407_at | - | - |
| Mouse Expression Set 430 | 1442407_PM_at | - | - |
| Murine Genome U74v2 Set | 168124_at | - | - |
| Murine Genome U74v2 Set | 168124_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 417460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243408 | - | - |

Zkscan7
Genomic Map