Assay Details
| Gene Symbol: Slc6a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028878.3 | NP_083154.1 | 7 - 8 | 1079 | 66 | |
| GenBank mRNA | AJ633679.1 | - | 7 - 8 | 1088 | 66 | |
| AK014544.1 | - | 7 - 8 | 1080 | 66 | ||
| AK028784.1 | - | 7 - 8 | 1080 | 66 | ||
| AK128923.1 | - | 6 - 7 | 1422 | 66 | ||
| AK162399.1 | - | 6 - 7 | 824 | 66 | ||
| BC107411.1 | - | 7 - 8 | 1080 | 40059042 | 66 | |
| BC107412.2 | - | 7 - 8 | 1084 | 40059050 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a19
Genomic Map