Assay Details
| Gene Symbol: Slc6a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028878.3 | NP_083154.1 | 10 - 11 | 1601 | 66 | |
| GenBank mRNA | AB096860.1 | - | 8 - 9 | 1284 | 66 | |
| AJ633679.1 | - | 10 - 11 | 1610 | 66 | ||
| AK014544.1 | - | 10 - 11 | 1602 | 66 | ||
| AK028784.1 | - | 10 - 11 | 1602 | 66 | ||
| AK128923.1 | - | 9 - 10 | 1944 | 66 | ||
| AK162399.1 | - | 9 - 10 | 1346 | 66 | ||
| BC047519.1 | - | 1 - 2 | 103 | 66 | ||
| BC107411.1 | - | 10 - 11 | 1602 | 40059042 | 66 | |
| BC107412.2 | - | 10 - 11 | 1606 | 40059050 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a19
Genomic Map