Assay Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144904.2 | NP_659153.2 | 10 - 11 | 1309 | 133 | |
| NM_178164.3 | NP_835458.1 | 9 - 10 | 1284 | 133 | ||
| GenBank mRNA | AK033732.1 | - | 9 - 10 | 1209 | 133 | |
| AK042920.1 | - | 7 - 8 | 1216 | 133 | ||
| AK088560.1 | - | 9 - 10 | 1216 | 133 | ||
| BC006638.1 | - | 10 - 11 | 1219 | 3969488 | 133 | |
| BC057641.1 | - | 9 - 10 | 1189 | 4936557 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp3
Genomic Map