Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252443.1 | NP_001239372.1 | 3 - 4 | 578 | 87 | |
| GenBank mRNA | AK088864.1 | - | 3 - 4 | 396 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfg
Genomic Map