Assay Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011571.3 | NP_035701.3 | 7 - 8 | 1939 | 74 | |
| GenBank mRNA | AK144302.1 | - | 7 - 8 | 1522 | 74 | |
| BC099699.1 | - | 7 - 8 | 1939 | 6821934 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tesk1
Genomic Map