Assay Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011571.3 | NP_035701.3 | 8 - 9 | 2030 | 73 | |
| GenBank mRNA | AB003494.1 | - | 8 - 9 | 2368 | 73 | |
| AK144302.1 | - | 8 - 9 | 1613 | 73 | ||
| BC099699.1 | - | 8 - 9 | 2030 | 6821934 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tesk1
Genomic Map