Assay Details
| Gene Symbol: Creb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013497.3 | NP_038525.2 | 3 - 4 | 552 | 147 | |
| GenBank mRNA | AK090077.1 | - | 3 - 4 | 522 | 147 | |
| AK132785.1 | - | 3 - 4 | 562 | 147 | ||
| AK144208.1 | - | 3 - 4 | 555 | 147 | ||
| BC002094.1 | - | 3 - 4 | 518 | 3490360 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3
Genomic Map