Assay Details
| Gene Symbol: Creb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013497.3 | NP_038525.2 | 5 - 6 | 752 | 72 | |
| GenBank mRNA | AK007665.1 | - | 2 - 3 | 181 | 72 | |
| AK132785.1 | - | 5 - 6 | 762 | 72 | ||
| AK144208.1 | - | 5 - 6 | 755 | 72 | ||
| BC002094.1 | - | 5 - 6 | 718 | 3490360 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3
Genomic Map