Assay Details
| Gene Symbol: Creb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013497.3 | NP_038525.2 | 6 - 7 | 819 | 77 | |
| GenBank mRNA | AK007665.1 | - | 3 - 4 | 248 | 77 | |
| AK090077.1 | - | 6 - 7 | 790 | 77 | ||
| AK132785.1 | - | 6 - 7 | 829 | 77 | ||
| AK144208.1 | - | 6 - 7 | 822 | 77 | ||
| BC002094.1 | - | 6 - 7 | 785 | 3490360 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3
Genomic Map