Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 12 - 13 | 1010 | 93 | |
| GenBank mRNA | AK029992.1 | - | 13 - 14 | 1240 | 93 | |
| AK076708.1 | - | 12 - 13 | 1011 | 93 | ||
| EF377342.1 | - | 12 - 13 | 999 | 93 | ||
More Information
Set Membership: |
|

Atp8b5
Genomic Map