Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 20 - 21 | 2205 | 112 | |
| GenBank mRNA | AK029992.1 | - | 21 - 22 | 2436 | 112 | |
| EF377342.1 | - | 20 - 21 | 2194 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b5
Genomic Map