Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 23 - 24 | 2643 | 103 | |
| GenBank mRNA | AK029992.1 | - | 24 - 25 | 2874 | 103 | |
More Information
Set Membership: |
|

Atp8b5
Genomic Map