Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 25 - 26 | 2953 | 75 | |
| GenBank mRNA | AK029992.1 | - | 26 - 27 | 3184 | 75 | |
| EF377342.1 | - | 25 - 26 | 2942 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b5
Genomic Map