Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 5 - 6 | 377 | 77 | |
| GenBank mRNA | AK029992.1 | - | 6 - 7 | 607 | 77 | |
| AK076708.1 | - | 5 - 6 | 378 | 77 | ||
| EF377342.1 | - | 5 - 6 | 366 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b5
Genomic Map