Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 7 - 8 | 530 | 131 | |
| GenBank mRNA | AK029992.1 | - | 8 - 9 | 760 | 131 | |
| AK076708.1 | - | 7 - 8 | 531 | 131 | ||
| AK078567.1 | - | 7 - 8 | 499 | 131 | ||
| EF377342.1 | - | 7 - 8 | 519 | 131 | ||
More Information
Set Membership: |
|

Atp8b5
Genomic Map