Assay Details
| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172692.3 | NP_766280.2 | 7 - 8 | 1775 | 90 | |
| GenBank mRNA | AK089192.1 | - | 7 - 8 | 1461 | 90 | |
| AK158174.1 | - | 7 - 8 | 1775 | 90 | ||
| AK173213.1 | - | 7 - 8 | 1761 | 90 | ||
| BC031397.1 | - | 6 - 7 | 1610 | 90 | ||
| BC034105.1 | - | 6 - 7 | 1155 | 90 | ||
| BC056935.1 | - | 7 - 8 | 1298 | 5703478 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gba2
Genomic Map