Assay Details
| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172692.3 | NP_766280.2 | 8 - 9 | 1894 | 93 | |
| GenBank mRNA | AK089192.1 | - | 8 - 9 | 1580 | 93 | |
| AK173213.1 | - | 8 - 9 | 1880 | 93 | ||
| BC056935.1 | - | 8 - 9 | 1417 | 5703478 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gba2
Genomic Map