Assay Details
| Gene Symbol: Fancg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163233.1 | NP_001156705.1 | 10 - 11 | 1627 | 65 | |
| NM_053081.2 | NP_444311.1 | 10 - 11 | 1648 | 65 | ||
| GenBank mRNA | AF112439.1 | - | 10 - 11 | 1457 | 65 | |
| AY049715.1 | - | 10 - 11 | 1448 | 65 | ||
| BC050890.1 | - | 10 - 11 | 1905 | 65 | ||
| BC139763.1 | - | 10 - 11 | 1430 | 40110334 | 65 | |
| BC141282.1 | - | 10 - 11 | 1464 | 9055894 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fancg
Genomic Map