Assay Details
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025900.2 | NP_080176.2 | 8 - 9 | 1070 | 115 | |
GenBank mRNA | AK007546.1 | - | 5 - 6 | 619 | 115 | |
AK033451.1 | - | 8 - 9 | 960 | 115 | ||
AK141790.1 | - | 2 - 3 | 422 | 115 | ||
BC055451.1 | - | 8 - 9 | 1016 | 4164972 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
