Assay Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178848.3 | NP_849179.1 | 6 - 7 | 793 | 54 | |
| GenBank mRNA | AK002609.1 | - | 7 - 8 | 822 | 54 | |
| AK005346.1 | - | 6 - 7 | 806 | 54 | ||
| BC031770.1 | - | 6 - 7 | 793 | 4014119 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sirt5
Genomic Map