Assay Details
| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183392.2 | NP_899248.1 | 10 - 11 | 1419 | 94 | |
| GenBank mRNA | AK088855.1 | - | 10 - 11 | 1344 | 94 | |
| AK133841.1 | - | 10 - 11 | 1417 | 94 | ||
| AK150182.1 | - | 10 - 11 | 1344 | 94 | ||
| AK150841.1 | - | 10 - 11 | 1306 | 94 | ||
| AK151896.1 | - | 10 - 11 | 1306 | 94 | ||
| AK152136.1 | - | 10 - 11 | 1306 | 94 | ||
| AK162037.1 | - | 10 - 11 | 1351 | 94 | ||
| BC044303.1 | - | 10 - 11 | 1344 | 94 | ||
| BC068114.1 | - | 10 - 11 | 1304 | 5707888 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup54
Genomic Map