Assay Details
| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183392.2 | NP_899248.1 | 4 - 5 | 658 | 130 | |
| GenBank mRNA | AK087827.1 | - | 4 - 5 | 552 | 130 | |
| AK088855.1 | - | 4 - 5 | 583 | 130 | ||
| AK133841.1 | - | 4 - 5 | 656 | 130 | ||
| AK150182.1 | - | 4 - 5 | 583 | 130 | ||
| AK150841.1 | - | 4 - 5 | 544 | 130 | ||
| AK151896.1 | - | 4 - 5 | 544 | 130 | ||
| AK152136.1 | - | 4 - 5 | 544 | 130 | ||
| AK162037.1 | - | 4 - 5 | 590 | 130 | ||
| BC044303.1 | - | 4 - 5 | 586 | 130 | ||
| BC068114.1 | - | 4 - 5 | 543 | 5707888 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup54
Genomic Map