Assay Details
| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183392.2 | NP_899248.1 | 7 - 8 | 1094 | 64 | |
| GenBank mRNA | AK087827.1 | - | 7 - 8 | 988 | 64 | |
| AK088855.1 | - | 7 - 8 | 1019 | 64 | ||
| AK133841.1 | - | 7 - 8 | 1092 | 64 | ||
| AK150182.1 | - | 7 - 8 | 1019 | 64 | ||
| AK150841.1 | - | 7 - 8 | 980 | 64 | ||
| AK151896.1 | - | 7 - 8 | 980 | 64 | ||
| AK152136.1 | - | 7 - 8 | 980 | 64 | ||
| AK162037.1 | - | 7 - 8 | 1026 | 64 | ||
| BC044303.1 | - | 7 - 8 | 1022 | 64 | ||
| BC068114.1 | - | 7 - 8 | 979 | 5707888 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup54
Genomic Map