Assay Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271397.1 | NP_001258326.1 | 11 - 12 | 3388 | 68 | |
| NR_073167.1 | 11 - 12 | 3569 | 68 | |||
| NR_073168.1 | 11 - 12 | 3384 | 68 | |||
| GenBank mRNA | AK017551.1 | - | 4 - 5 | 464 | 68 | |
| AK029961.1 | - | 11 - 12 | 3101 | 68 | ||
| AK147172.1 | - | 11 - 12 | 3124 | 68 | ||
| BC049166.1 | - | 11 - 12 | 2978 | 68 | ||
| BC058699.1 | - | 11 - 12 | 3544 | 68 | ||
| BC125575.1 | - | 11 - 12 | 3114 | 40130090 | 68 | |
| BC125579.1 | - | 11 - 12 | 3114 | 40130094 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nol8
Genomic Map