Assay Details
| Gene Symbol: Ccr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007720.2 | NP_031746.1 | 1 - 2 | 57 | 79 | |
| GenBank mRNA | Z98205.1 | - | 1 - 2 | 57 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_007720.2, Z98205.1 transcripts.
Set Membership: |
|

Ccr8
Genomic Map